ENA181911290 Enamine

N-[5-(4-hydroxyphenyl)-1,3-thiazol-2-yl]acetamide

  • MDL number MFCD11099631
  • Cas Number : 1087792-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39087

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39087
Availability details for
Quantity
  • Availability
  • Add to Cart