ENA181911291 Enamine

3-[(trifluoromethyl)sulfonyl]propanoic acid

  • MDL number MFCD11099632
  • Cas Number : 1087784-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104...106
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39090

Safety Information

msds https://www.enaminestore.com/catalog/EN300-39090
Availability details for
Quantity
  • Availability
  • Add to Cart