ENA181911297 Enamine

3-chloro-4-phenyl-4H-1,2,4-triazole

  • MDL number MFCD09995764
  • Cas Number : 90002-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-39131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39131
Availability details for
Quantity
  • Availability
  • Add to Cart