ENA181911302 Enamine

2-[4-(1-aminoethyl)-2-methoxyphenoxy]acetic acid hydrochloride

  • MDL number MFCD11099643
  • Cas Number : 1170395-91-1
  • Molecular Weight : 261.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-39152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39152
Availability details for
Quantity
  • Availability
  • Add to Cart