ENA181911314 Enamine

2-(butylamino)acetamide hydrochloride

  • MDL number MFCD11099657
  • Cas Number : 91400-34-9
  • Molecular Weight : 166.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39200
Availability details for
Quantity
  • Availability
  • Add to Cart