ENA181911316 Enamine

N-[2-(diethylamino)ethyl]piperidine-4-carboxamide hydrochloride

  • MDL number MFCD11099659
  • Cas Number : 1171023-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39207
Availability details for
Quantity
  • Availability
  • Add to Cart