ENA181911317 Enamine

propyl(2,2,2-trifluoroethyl)amine hydrochloride

  • MDL number MFCD11099663
  • Cas Number : 405879-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-39245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-39245
Availability details for
Quantity
  • Availability
  • Add to Cart