ENA181911375 Enamine

2-fluoro-6-(piperazin-1-yl)benzonitrile hydrochloride

  • MDL number MFCD11099448
  • Cas Number : 1172753-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36877
Availability details for
Quantity
  • Availability
  • Add to Cart