ENA181911382 Enamine

N,N-dimethyl-2-(4-oxopiperidin-1-yl)acetamide

  • MDL number MFCD09951130
  • Cas Number : 1017026-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36886
Availability details for
Quantity
  • Availability
  • Add to Cart