ENA181911387 Enamine

({[(3-methylphenyl)amino]carbonyl}amino)acetic acid

  • MDL number MFCD08444862
  • Cas Number : 133115-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182...184
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36892

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36892
Availability details for
Quantity
  • Availability
  • Add to Cart