ENA181911401 Enamine

N-(4-amino-2-methoxyphenyl)pyridine-3-carboxamide

  • MDL number MFCD02585685
  • Cas Number : 552814-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36913

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36913
Availability details for
Quantity
  • Availability
  • Add to Cart