ENA181911402 Enamine

3-chloro-4-[(pyridin-3-yl)methoxy]aniline

  • MDL number MFCD08699390
  • Cas Number : 848134-24-7
  • Molecular Weight : 234.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-36914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36914
Availability details for
Quantity
  • Availability
  • Add to Cart