ENA181911404 Enamine

methyl 2-amino-5-(morpholin-4-yl)benzoate

  • MDL number MFCD11099382
  • Cas Number : 134050-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27630
Availability details for
Quantity
  • Availability
  • Add to Cart