ENA181911405 Enamine

3-[3-(propan-2-yl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD11048465
  • Cas Number : 947013-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27664
Availability details for
Quantity
  • Availability
  • Add to Cart