ENA181911407 Enamine

ethyl 3-amino-4-cyano-1-(2-methoxyethyl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD08444749
  • Cas Number : 923238-26-0
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36920

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36920
Availability details for
Quantity
  • Availability
  • Add to Cart