ENA181911423 Enamine

1-(4-acetylphenyl)-3-(propan-2-yl)urea

  • MDL number MFCD06115045
  • Cas Number : 72531-19-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36937

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36937
Availability details for
Quantity
  • Availability
  • Add to Cart