ENA181911431 Enamine

(3-aminophenyl)urea

  • MDL number MFCD00035935
  • Cas Number : 25711-72-2
  • Molecular Weight : 151.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 128-130
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-28242

Safety Information

signalword Warning
pcodes P261,P264,P271,P272,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P333+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-28242
pictograms GHS07
hstatements H315,H317,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart