ENA181911438 Enamine

2-phenoxypyridin-3-amine

  • MDL number MFCD09738154
  • Cas Number : 28664-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36953
Availability details for
Quantity
  • Availability
  • Add to Cart