ENA181911444 Enamine

9H-fluorene-2-carboxylic acid

  • MDL number MFCD00034611
  • Cas Number : 7507-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 276-278
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-36960

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36960
Availability details for
Quantity
  • Availability
  • Add to Cart