ENA181911445 Enamine

3-(1H-1,2,4-triazol-1-yl)propan-1-amine

  • MDL number MFCD03727267
  • Cas Number : 69807-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-36963

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36963
Availability details for
Quantity
  • Availability
  • Add to Cart