ENA181911447 Enamine

1-(2-hydroxyethyl)-6-oxo-1,4,5,6-tetrahydropyridazine-3-carboxylic acid

  • MDL number MFCD09050274
  • Cas Number : 349134-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36965

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36965
Availability details for
Quantity
  • Availability
  • Add to Cart