ENA181911452 Enamine

N-cyclopropyl-2-hydroxybenzamide

  • MDL number MFCD02934388
  • Cas Number : 440111-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36970

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36970
Availability details for
Quantity
  • Availability
  • Add to Cart