ENA181911453 Enamine

4-methoxy-3-[(oxolan-2-yl)methoxy]aniline

  • MDL number MFCD09737325
  • Cas Number : 954254-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36971
Availability details for
Quantity
  • Availability
  • Add to Cart