ENA181911454 Enamine

ethyl (chlorosulfonyl)acetate

  • MDL number MFCD08458866
  • Cas Number : 55896-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36972

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-36972
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart