ENA181911459 Enamine

5-amino-3-methoxy-1H-pyrazole-4-carbonitrile

  • MDL number MFCD09702270
  • Cas Number : 72760-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36977

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36977
Availability details for
Quantity
  • Availability
  • Add to Cart