ENA181911462 Enamine

3-ethylpiperidine

  • MDL number MFCD01700577
  • Cas Number : 13603-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36982

Safety Information

msds https://www.enaminestore.com/catalog/EN300-36982
Availability details for
Quantity
  • Availability
  • Add to Cart