ENA181911473 Enamine

5-(6-methylpyridin-2-yl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD09811080
  • Cas Number : 1016520-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-36995

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-36995
Availability details for
Quantity
  • Availability
  • Add to Cart