ENA181911477 Enamine

2-(2-oxo-1,3-oxazolidin-3-yl)ethanethioamide

  • MDL number MFCD01755554
  • Cas Number : 172514-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37011
Availability details for
Quantity
  • Availability
  • Add to Cart