ENA181911486 Enamine

methyl (2E)-4-(4-methoxyphenyl)-4-oxobut-2-enoate

  • MDL number MFCD11099480
  • Cas Number : 17615-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37026
Availability details for
Quantity
  • Availability
  • Add to Cart