ENA181911506 Enamine

4-(ethylsulfamoyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09052902
  • Cas Number : 937627-33-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37074
Availability details for
Quantity
  • Availability
  • Add to Cart