ENA181911520 Enamine

3-amino-2-[(2-methylphenyl)methyl]propan-1-ol

  • MDL number MFCD09894876
  • Cas Number : 1017371-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37090

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37090
Availability details for
Quantity
  • Availability
  • Add to Cart