ENA181911522 Enamine

3-amino-2-[(3-methylphenyl)methyl]propan-1-ol

  • MDL number MFCD09894877
  • Cas Number : 1017184-58-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37092
Availability details for
Quantity
  • Availability
  • Add to Cart