ENA181911532 Enamine

2-(cyclopropylamino)ethan-1-ol

  • MDL number MFCD09815646
  • Cas Number : 35265-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37102
Availability details for
Quantity
  • Availability
  • Add to Cart