ENA181911538 Enamine

3-(2,3,5-trimethylphenoxy)propanenitrile

  • MDL number MFCD10695617
  • Cas Number : 3649-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37112
Availability details for
Quantity
  • Availability
  • Add to Cart