ENA181911540 Enamine

quinolin-4-amine hydrochloride

  • MDL number MFCD11099495
  • Cas Number : 35654-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37114

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37114
Availability details for
Quantity
  • Availability
  • Add to Cart