ENA181911543 Enamine

2-(3,4-dimethylphenoxy)isonicotinonitrile

  • MDL number MFCD09944940
  • Cas Number : 1016822-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63...65
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37118
Availability details for
Quantity
  • Availability
  • Add to Cart