ENA181911545 Enamine

2-[4-(carbamoylamino)phenyl]acetic acid

  • MDL number MFCD09945533
  • Cas Number : 69100-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-22411

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22411
Availability details for
Quantity
  • Availability
  • Add to Cart