ENA181911551 Enamine

4-(methylsulfamoyl)-1H-pyrrole-2-carboxylic acid

  • MDL number MFCD09053121
  • Cas Number : 937629-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37128
Availability details for
Quantity
  • Availability
  • Add to Cart