ENA181911560 Enamine

methyl (2E)-4-oxo-4-phenylbut-2-enoate

  • MDL number MFCD01316720
  • Cas Number : 14274-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37144

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37144
Availability details for
Quantity
  • Availability
  • Add to Cart