ENA181911568 Enamine

3-methyl-1-phenyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09049487
  • Cas Number : 926253-76-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37159
Availability details for
Quantity
  • Availability
  • Add to Cart