ENA181911579 Enamine

4-(chloromethyl)-2-[(4-chlorophenyl)methyl]-1,3-thiazole

  • MDL number MFCD09050363
  • Cas Number : 73765-41-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37183

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37183
Availability details for
Quantity
  • Availability
  • Add to Cart