ENA181911581 Enamine

(1E)-1-(2,4-dimethyl-1,3-thiazol-5-yl)ethanone oxime

  • MDL number MFCD10044431
  • Cas Number : 40236-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37185

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37185
Availability details for
Quantity
  • Availability
  • Add to Cart