ENA181911584 Enamine

2-(4-aminophenyl)-1-[4-(pyrimidin-2-yl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD09816665
  • Cas Number : 1016694-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37188

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37188
Availability details for
Quantity
  • Availability
  • Add to Cart