ENA181911585 Enamine

[2-(4-methoxyphenyl)ethyl]thiourea

  • MDL number MFCD11099391
  • Cas Number : 81050-04-6
  • Molecular Weight : 210.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-28858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28858
Availability details for
Quantity
  • Availability
  • Add to Cart