ENA181911588 Enamine

3-amino-2-methylbenzamide

  • MDL number MFCD09028329
  • Cas Number : 374889-30-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37193

Safety Information

msds https://www.enaminestore.com/catalog/EN300-37193
Availability details for
Quantity
  • Availability
  • Add to Cart