ENA181911593 Enamine

N-[(2-aminophenyl)methyl]-N-ethylacetamide

  • MDL number MFCD11099512
  • Cas Number : 1087792-25-3
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-37198

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37198
Availability details for
Quantity
  • Availability
  • Add to Cart