ENA181911594 Enamine

3-(aminomethyl)-N-methylbenzene-1-sulfonamide

  • MDL number MFCD09732338
  • Cas Number : 808761-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-37199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37199
Availability details for
Quantity
  • Availability
  • Add to Cart