ENA181911595 Enamine

8-fluoro-6-methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09739536
  • Cas Number : 954260-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-37200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37200
Availability details for
Quantity
  • Availability
  • Add to Cart