ENA181911596 Enamine

1-[3-methoxy-4-(2,2,2-trifluoroethoxy)phenyl]ethan-1-amine

  • MDL number MFCD09735712
  • Cas Number : 953750-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-37201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37201
Availability details for
Quantity
  • Availability
  • Add to Cart