ENA181911602 Enamine

1-(3-cyanophenyl)-3-cyclopropylurea

  • MDL number MFCD11099513
  • Cas Number : 1087784-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-37208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-37208
Availability details for
Quantity
  • Availability
  • Add to Cart